Learn More
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Especificaciones
| Chemical Name or Material | Silver nitrate |
| Name Note | 0.1 N Standard Solution |
| CAS | 7761-88-8, 7732-18-5 |
| Color | Colorless |
| Physical Form | Liquid |
| Quantity | 5 L |
| Molecular Formula | AgNO3 |
| Linear Formula | AgNO3 |
| MDL Number | MFCD00003414 |
| Fieser | 01,1008; 02,366; 03,252; 04,429; 05,582; 07,321; 09,411; 10,350; 11,268; 12,257; 14,350; 15,39 |
| Mostrar más |
Seguridad y manipulación
- Silver nitrate, 0.1N standard solution
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 2
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
- H290-May be corrosive to metals.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H410-Very toxic to aquatic life with long lasting effects.
- P234-Keep only in original container.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P403-Store in a well-ventilated place.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: Silver nitrate
RUO – Research Use Only
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.