missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Silver Nitrate, 99.8+%, Extra Pure, SLR, Fisher Chemical™
Shop alle Fisher Chemical produkter
Klik for at se tilgængelige muligheder
Quantity:
25 g
100 g
250 g
500 g
Pakningsstørrelse:
100g
250g
25g
500g
Specifications
Specifications
| Boiling Point | 444°C |
| Melting Point | 212°C |
| Color | White |
| pH | 5.4 to 6.4 |
| Physical Form | Solid |
| Quantity | 500 g |
| IUPAC Name | silver(1+) nitrate |
| Formula Weight | 169.87g/mol |
| Packaging | Glass Jar |
Safety and Handling
Product Identifier
- Silver nitrate
- Danger
- Oxidising solid Category 2
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H272-May intensify fire; oxidiser.
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H410-Very toxic to aquatic life with long lasting effects.
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction