missing translation for 'onlineSavingsMsg'
Learn More

Silver nitrate, 99+%, ACS reagent

Product Code. 10617411 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
250g
500g
50g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10617411 500 g Glass bottle 500g
10422981 50 g Glass bottle 50g
10599092 100 g Glass bottle 100g
10148280 250 g Glass bottle 250g
11984341 2.5 kg Glass bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10617411 Supplier Thermo Scientific Acros Supplier No. 419355000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 7761-88-8 | AgNO3 | 169.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Boiling Point 444.0°C
Melting Point 212.0°C
Color White
Physical Form Crystalline Powder and/or Crystals
Quantity 500 g
Assay Percent Range 99+%
Linear Formula AgNO3
Fieser 01,1008; 02,366; 03,252; 04,429; 05,582; 07,321; 09,411; 10,350; 11,268; 12,257; 14,350; 15,39
Merck Index 15, 8657
Solubility Information Solubility in water: 219 g/100mL (20°C). Other solubilities: soluble in alcohol, slightly soluble in acetone and ether
Formula Weight 169.87
Percent Purity ≥99%
Grade ACS Reagent
Packaging Glass bottle
Decomposition Information >444°C
Density 4.35
Chemical Name or Material Silver nitrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.