missing translation for 'onlineSavingsMsg'
Learn More

Sodium chloride, crystalline powder PDV, 99+%

Product Code. 11460990
Change view
Click to view available options
Quantity:
1009 Ea.
Packaging:
Glass jar
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11460990 1009 Ea. Glass jar 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11460990 Supplier Thermo Scientific Alfa Aesar Supplier No. A13854.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7761-88-8 | AgNO3 | 169.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

  • Employed in Silver plating, photography, manufacturing of other silver compounds, mirrors, coloring porcelain, and staining in histology.
  • Finds use as a stain in scanning electron microscopy, and a key ingredient in several silver-based antiseptic and antibacterial pharmaceutical compositions.
  • Used to separate mixtures of alkenes by selective absorption, and for deprotection and oxidation reactions in organic synthesis.
  • Widely used as an analytical reagent owing to its immediate reaction with halide ions forming insoluble precipitate of silver(I) halides
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 444°C
Melting Point 212°C
CAS 7761-88-8
Physical Form Crystalline
Quantity 1009 Ea.
Assay Percent Range 99+%
Molecular Formula AgNO3
MDL Number MFCD00003414
UN Number UN1493
Merck Index 14,8518
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
Solubility Information Freely soluble in water, alcohol and aqueous ammonia. Slightly soluble in ether.
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
SMILES [Ag+].[O-][N+]([O-])=O
IUPAC Name silver;nitrate
Molecular Weight (g/mol) 169.87
PubChem CID 24470
ChEBI CHEBI:32130
Formula Weight 169.87
Packaging Glass jar
Sensitivity Light Sensitive
Appearance White
Decomposition Information > 444°C
Density 4.352
Chemical Name or Material Silver nitrate
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Oxidising solid Category 2
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H360D-May damage the unborn child.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.