missing translation for 'onlineSavingsMsg'
Learn More

Silver nitrate, ACS, 99.9+% (metals basis)

Product Code. p-4407818
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11361229 5 g 5g
11371229 25 g 25g
11381229 100 g 100g
11391229 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11361229 Supplier Thermo Scientific Alfa Aesar Supplier No. 011414.06

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Silver nitrate, AgNO3, CAS Number-7761-88-8, silvernitrate, silver i nitrate, argenti nitras, nitric acid silver i salt, nitrate d'argent, lunar caustic, argerol, silver mononitrate, silver nitrate, silbernitrat, 100g, >99.9% (metals basis), CHEBI:32130, 4.352 g/mL | CAS: 7761-88-8 | AgNO3 | 169.87 g/mol

Silver nitrate is employed in Silver plating, photography, manufacturing of other silver compounds, mirrors, coloring porcelain, and staining in histology. It finds use as a stain in scanning electron microscopy, and a key ingredient in several silver-based antiseptic and antibacterial pharmaceutical compositions. Silver(I) nitrate is used to separate mixtures of alkenes by selective absorption, and for deprotection and oxidation reactions in organic synthesis. It is widely used as an analytical reagent owing to its immediate reaction with halide ions forming insoluble precipitate of silver(I) halides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O

Especificaciones

Melting Point 212°C
Physical Form Crystalline
Quantity 5 g
Assay Percent Range >99.9% (metals basis)
Solubility Information Freely soluble in water,alcohol and aqueous ammonia. Slightly soluble in ether.
Formula Weight 169.87
Grade ACS Reagent
Sensitivity Light sensitive
Density 4.352 g/mL
Chemical Name or Material Silver nitrate, For AAS analysis
compliance-icons compliance-icons compliance-icons
Nombre del producto
  • Silver nitrate
Palabra de advertencia
  • Danger
Clasificación
  • Oxidising solid Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Indicaciones de peligro
  • H272-May intensify fire; oxidiser.
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H410-Very toxic to aquatic life with long lasting effects.
Consejos de prudencia
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.