missing translation for 'onlineSavingsMsg'
Learn More

Silver Nitrate (Crystalline/Certified ACS), Fisher Chemical™

Product Code. 10265063
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10265063 500 g Glass Bottle 500g
10346843 25 g Glass Bottle 25g
10284773 100 g Glass Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10265063 Supplier Fisher Chemical Supplier No. S181500
 Restricted Item
Estimated Shipment: 16-03-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Boiling Point 444°C
Melting Point 212°C
Color White
pH 5.4 to 6.4
Physical Form Solid
Quantity 500 g
Assay Percent Range Assay: ≥99.7%
Formula Weight 169.87
Percent Purity ≥99.7%
Grade Certified ACS
Identification Pass Test
Packaging Glass Bottle
Chemical Name or Material Silver Nitrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.