missing translation for 'onlineSavingsMsg'
Learn More

Silver nitrate, Premion™, 99.995% (metals basis), Ag 63% min

Product Code. 11394738
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11394738 5 g 5g
11304748 25 g 25g
11384738 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11394738 Supplier Thermo Scientific Alfa Aesar Supplier No. 043087.06
 Restricted Item
Request Bulk or Custom Format

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7761-88-8 | AgNO3 | 169.87 g/mol

Silver nitrate is employed in Silver plating, photography, manufacturing of other silver compounds, mirrors, coloring porcelain, and staining in histology. It finds use as a stain in scanning electron microscopy, and a key ingredient in several silver-based antiseptic and antibacterial pharmaceutical compositions. Silver(I) nitrate can be used to separate mixtures of alkenes by selective absorption, and for deprotection and oxidation reactions in organic synthesis. It is widely used as an analytical reagent owing to its immediate reaction with halide ions forming insoluble precipitate of silver(I) halides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Melting Point 212°C
Color White
Physical Form Crystalline
Quantity 5 g
UN Number UN1479
Merck Index 14,8518
Solubility Information Freely soluble in water,alcohol and aqueous ammonia. Slightly soluble in ether.
Formula Weight 169.87
Percent Purity 99.995%
Grade Premion
Concentration or Composition (by Analyte or Components) Ag 63% min
Odor Odorless
Sensitivity Light sensitive
Density 4.352 g/mL
Assay (metals basis)
Chemical Name or Material Silver nitrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.