missing translation for 'onlineSavingsMsg'
Learn More

Silver nitrate solution, Volumetric, Reag. Ph. Eur., 0.1 M AgNO3 (0.1N), Honeywell Fluka™

Product Code. 15615020 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
10 L
6 x 1 L
missing translation for 'unitSize'
1 L
10 L
2.5 L
5 L
500mL
Pack of 6
Código de producto. Quantity unitSize
15615020 500 mL 500mL
15695010 1 L 1 L
15605020 2.5 L 2.5 L
15635020 5 L 5 L
15645020 6 x 1 L Pack of 6
15685010 10 L 10 L
6 missing translation for 'options'
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15615020

missing translation for 'mfr': Honeywell-Fluka 35375500ML

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

Volumetric, Reag. Ph. Eur., 0.1 M AgNO3 (0.1N)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
SMILES [Ag+].[O-][N+]([O-])=O

Especificaciones

Quantity 500 mL
UN Number UN3082
IUPAC Name silver(1+) nitrate
Formula Weight 169.87g/mol
Concentration or Composition (by Analyte or Components) 0.1M
Chemical Name or Material Silver nitrate solution
compliance-icons compliance-icons
Nombre del producto
  • Silver nitrate solution
Palabra de advertencia
  • Warning
Clasificación
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Indicaciones de peligro
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
Consejos de prudencia
  • P234-Keep only in original container.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Otros peligros
  • MIXTURE LIST-Contains : Silver nitrate solution
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.