missing translation for 'onlineSavingsMsg'
Learn More

Silver nitrate, TraceSELECT™, ≥99.998% (metals basis), Honeywell™ Flula™

Product Code. 15645490 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
25 g
Packaging:
Glass Bottle
Unit Size:
25g
This item is not returnable. View return policy

Product Code. 15645490

Brand: Honeywell-Fluka 4243425G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

TraceSELECT™, for trace analysis,≥99.998% (metals basis)

TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions

 


 

TRUSTED_SUSTAINABILITY

Specifications

CAS 7761-88-8
Quantity 25 g
Molecular Formula AgNO3
Linear Formula AgNO3
MDL Number MFCD00003414
UN Number UN1493
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
SMILES [Ag+].[O-][N+]([O-])=O
IUPAC Name silver(1+) nitrate
Molecular Weight (g/mol) 169.87
PubChem CID 24470
ChEBI CHEBI:32130
Formula Weight 169.87g/mol
Percent Purity ≥99.998% (metals basis)
Grade TraceSELECT
Packaging Glass Bottle
Density Vapor Density: 5.8 (vs air)
Chemical Name or Material Silver nitrate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.