missing translation for 'onlineSavingsMsg'
Learn More

Silver phosphate, 98%

Product Code. 10157610 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10157610 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10157610 Supplier Thermo Scientific Acros Supplier No. 419370250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

CAS: 7784-09-0 | Ag3O4P | 418.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 849°C
CAS 7784-09-0
Color Green-Yellow to Yellow
Physical Form Powder
Quantity 25 g
Assay Percent Range 97.5% min
Molecular Formula Ag3O4P
Linear Formula Ag3PO4
MDL Number MFCD00014148
Merck Index 15, 8664
Synonym trisilver phosphate, trisilver 1+ phosphate, unii-zl6t4y1xp8, zl6t4y1xp8, silver orthophosphate, trisilver orthophosphate, silver phosphate ag3po4, silberphosphat, trisilver 1+ orthophosphate, silver orthophosphate ag3po4
Solubility Information Solubility in water: negligible. Other solubilities: slightly sol in dil acetic acid,freely sol in dil nitric acid,in ammonia,,ammonium carbonate,also in alkali cyanides and,thiosulfates
InChI Key FJOLTQXXWSRAIX-UHFFFAOYSA-K
SMILES [Ag+].[Ag+].[Ag+].[O-]P([O-])([O-])=O
IUPAC Name trisilver;phosphate
Molecular Weight (g/mol) 418.57
PubChem CID 159913
Formula Weight 418.57
Percent Purity 98%
Packaging Glass bottle
Chemical Name or Material Silver phosphate, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Silver phosphate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.