missing translation for 'onlineSavingsMsg'
Learn More

Silver Sulfate, Certified AR for Analysis, Fisher Chemical™

Product Code. 11448822 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Amber glass bottle
Glass Jar
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11448822 500 g Amber glass bottle 500g
10090340 25 g Glass Jar 25g
10774381 100 g Glass Jar 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Artikelnummer. 11448822 missing translation for 'mfr' Fisher Chemical missing translation for 'supplierNo' S/1400/53
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Chemische Identifikatoren

CAS 10294-26-5
Molecular Formula Ag2H2O4S
Molecular Weight (g/mol) 313.81
MDL Number 3407
InChI Key YPNVIBVEFVRZPJ-UHFFFAOYSA-N
Synonym silver sulfate, disilver sulfate, disilver 1+ sulfate, unii-8qg6hv4zpo, disilver 1+ sulphate, sulfuric acid, disilver 1+ salt, 8qg6hv4zpo, disilver 1+ ion sulfate, sulfuric acid, silver 1+ salt 1:2, sulfuric acid disilver i salt
PubChem CID 159865
IUPAC Name disilver(1+) sulfuric acid
SMILES [Ag+].[Ag+].OS(O)(=O)=O

Spezifikation

Boiling Point 1085°C
Melting Point 652°C
Color White
pH 5 to 6
Physical Form Solid
Quantity 500 g
Formula Weight 311.79g/mol
Packaging Amber glass bottle
compliance-icons compliance-icons
Produktidentifikator
  • Silver sulfate
Signalwort
  • Danger
Gefahrenkategorie
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
Gefahrenhinweis
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H318-Causes serious eye damage.
Sicherheitshinweis
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Name des Produkts
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.