missing translation for 'onlineSavingsMsg'
Learn More

Silver Sulfate, Extra Pure, SLR, Fisher Chemical™

Product Code. 10447650 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10447650 100 g 100g
10080340 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10447650 Supplier Fisher Chemical Supplier No. S/1360/48
Quantity Price
16 297.31 € / 100g
1 347.20 € / 100g
4 312.97 € / 100g

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ag2O4S, CAS Number-10294-26-5, disilver 1+ sulfate, sulfuric acid disilver i salt, disilver 1+ sulphate, disilver sulfate, unii-8qg6hv4zpo, sulfuric acid, silver 1+ salt 1:2, silver sulfate, sulfuric acid, disilver 1+ salt, disilver 1+ ion sulfate, 8qg6hv4zpo, 100g, 1085 deg.C, White, 311.79g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10294-26-5
Molecular Formula Ag2H2O4S
Molecular Weight (g/mol) 313.81
MDL Number 3407
InChI Key YPNVIBVEFVRZPJ-UHFFFAOYSA-N
Synonym silver sulfate, disilver sulfate, disilver 1+ sulfate, unii-8qg6hv4zpo, disilver 1+ sulphate, sulfuric acid, disilver 1+ salt, 8qg6hv4zpo, disilver 1+ ion sulfate, sulfuric acid, silver 1+ salt 1:2, sulfuric acid disilver i salt
PubChem CID 159865
IUPAC Name disilver(1+) sulfuric acid
SMILES [Ag+].[Ag+].OS(O)(=O)=O

Specifications

Boiling Point 1085°C
Melting Point 652°C
Color White
pH 5 to 6
Physical Form Solid
Quantity 100 g
Formula Weight 311.79g/mol
Packaging Glass Jar
compliance-icons compliance-icons
Product Identifier
  • Silver sulfate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.