missing translation for 'onlineSavingsMsg'
Learn More

Silver tubing, 6.35mm (0.25in) OD, 4.35mm (0.171in) ID, 99.9% (metals basis)

Product Code. 11327847
Change view
Click to view available options
Quantity:
10 cm
50 cm
4 x 50 cm
Unit Size:
10cm
50cm
Pack of 4
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11327847 10 cm 10cm
11307847 50 cm 50cm
11317847 4 x 50 cm Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11327847 Supplier Thermo Scientific Alfa Aesar Supplier No. 014259.BS
 Restricted Item
In Stock
Add to basket
Lägg i varukorgen
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

CAS: 7440-22-4 | Ag | 107.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 7440-22-4
Molecular Formula Ag
Molecular Weight (g/mol) 107.87
MDL Number MFCD00003397
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
Synonym argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
PubChem CID 23954
ChEBI CHEBI:9141
IUPAC Name silver
SMILES [Ag]

Specifikationer

Quantity 10 cm
Odor Characteristic
Weight ≈18g/10cm
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Silver tubing, OD: 6.35mm (0.25 in.) ID: 4.35mm (0.171 in.), Temper: as drawn

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.