missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver tungsten oxide, 99% (metals basis)
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
Description
Silver tungsten oxide is used in electronic and chemical industries. It is also used as a biochemical for proteomics research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Yellow |
| Quantity | 50 g |
| Odor | Odorless |
| Sensitivity | Light sensitive |
| Form | Powder |
| Linear Formula | Ag2WO4 |
| Solubility Information | Insoluble in water; Soluble in potassium cyanide, ammonium hydroxide and nitric acid |
| Formula Weight | 463.58 |
| Percent Purity | 99% |
| Chemical Name or Material | Silver tungsten oxide |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction