missing translation for 'onlineSavingsMsg'
Learn More

Silver two part conductive adhesive, EXP. XX/XX Resin and Hardener

Product Code. 11362028
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11362028 1 g 1g
11352028 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11362028 Supplier Thermo Scientific Alfa Aesar Supplier No. 012942.03
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
This item is not returnable. View return policy

A conductive epoxy which bonds well to metal, ceramics, plastics, glass and graphite

A conductive epoxy which bonds well to metal, ceramics, plastics, glass and graphite

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Limited shelf life, 60 days after shipment
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Silver two part conductive adhesive
Name Note EXP. x X/XX Resin and Hardener
Quantity 1 g
MDL Number MFCD00003397

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.