missing translation for 'onlineSavingsMsg'
Learn More

Silver two part conductive adhesive, EXP. XX/XX Resin and Hardener

Product Code. p-7049112
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 11362028

Brand: Thermo Scientific Alfa Aesar 012942.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A conductive epoxy which bonds well to metal, ceramics, plastics, glass and graphite

A conductive epoxy which bonds well to metal, ceramics, plastics, glass and graphite

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Limited shelf life, 60 days after shipment
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Silver two part conductive adhesive
Name Note EXP. x X/XX Resin and Hardener
Quantity 1 g
MDL Number MFCD00003397

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.