Learn More
Macherey-Nagel™ Silylating BSTFA
Product Code. 10407631 Shop All Macherey-Nagel Products
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
Pack of 5
Product Code. Quantity Unit Size
10407631 10 mL Pack of 5
1 options
Product Code. 10407631 Supplier Macherey-Nagel™ Supplier No. 701220.510
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Quantity 10 mL
compliance-icons compliance-icons compliance-icons
Product Identifier
  • BSTFA, 5x10 mL
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: N , O-bis-trimethylsilyl-trifluoroacetamide (BSTFA)