missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Silylating MBDSTFA

Product Code. 10388054 Shop All Macherey-Nagel Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10388054 1 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10388054 Supplier Macherey-Nagel™ Supplier No. 701440.101
  • ONLINE EXCLUSIVE PRICE
    397.00 €
    387.45 € / Pack of 10
    Save 9.55 €
    2% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Quantity 1 mL
compliance-icons compliance-icons
Product Identifier
  • MBDSTFA, 10x1 mL
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Supplemental information
  • MIXTURE LIST-Contain: MBDSTFA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.