Learn More
Macherey-Nagel™ Silylating TSIM
Product Code. 10269261 Shop All Macherey-Nagel Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
Pack of 20
Product Code. Quantity unitSize
10269261 1 mL Pack of 20
1 options
Product Code. 10269261 Supplier Macherey-Nagel™ Supplier No. 701310.201
Specifications
Item Description Silylating TSIM
Quantity 1 mL
compliance-icons compliance-icons compliance-icons
Product Identifier
  • TSIM, 20x1 mL
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Reproductive toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: N-trimethylsilyl-imidazole