missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Silylation Grade Solvents

Use specially manufactured and packaged Thermo Scientific™ Silylation Grade Solvents to meet the exacting needs of silylation and other sensitive derivatization reactions.

Brand:  Thermo Scientific™ TS-20672

Code : 16

Additional Details : CAS Number : 68-12-2 Weight : 1.81440kg Transport : UN number : 2265 Chem class : 3 Pack group : III


 View more versions of this product

Product Code. 11561941

  • 168.24 € / Each
Estimated Shipment: 16-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Use specially manufactured and packaged Thermo Scientific™ Silylation Grade Solvents to meet the exacting needs of silylation and other sensitive derivatization reactions. They are purified, dried, and packaged under nitrogen in convenient Thermo Scientific™ Hypo-Vial™ Sample Storage Vials, and are supplied with elastomer septa to enable immediate access to the sample without exposure to moisture and oxygen. Use polar solvents (acetonitrile, dimethylformamide, dimethylsulfoxide, pyridine, tetrahyrdrofuran) to facilitate reactions; nonpolar organic solvents may be used but will not accelerate the rate of reaction.

Recommended Use

Avoid water or alcohol, as TMS reagents react with active hydrogen.

Avoid enolizable ketones.

Use dimethylformamide for steroids and other large molecules.

Use dimethylsulfoxide to prepare TMS derivatives of tertiary alcohols and some compounds with low solubility in other silylation solvents.

Pyridine is an excellent solvent and reaction medium for MS reactions and is an HCI acceptor in reactions involving organochlorosilanes.

Other commonly used solvents include tetrahydrofuran and acetonitrile.

Specifications

Specifications

GC
Dimethylformamide (DMF); 50 mL
98 to 100%
Flammable, Harmful
Dimethylformamide (DMF)
4°C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Silylation Grade Solvents

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Reproductive toxicity Category 1

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H360-May damage fertility or the unborn child.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Dimethylformamide

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.