missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems Simple Reader™ ELISA Plate Reader

Product Code. 30245498
Change view
Click to view available options
Shaking:
None
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Shaking unitSize
30245498 None Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30245498 Supplier R&D Systems Supplier No. IA051123
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Empower every lab member with their own Simple Reader - a groundbreaking microplate reader featuring 96 individual detection units, ensuring each person has their own compact, reliable device for decentralized workflows.

Empower every lab member with their own Simple Reader - a groundbreaking microplate reader featuring 96 individual detection units, ensuring each person has their own compact, reliable device for decentralized workflows. Needing just a laptop, its plug-and-play functionality seamlessly transforms any location into your lab space, making fieldwork a breeze.

  • Compact Design: Take this personal ELISA plate reader on the go to any lab, anywhere.
  • 4 LED Light Sources: 450 nm, 490 nm, 540 nm, 595 nm
  • 96 Detection Units: Robust, reliable results every time.
  • Fast Read Time: Speed up data analysis by reading your plate in under 5 seconds.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Microplate Reader
Cuvettes No
Dimensions (L x W x H) 15.4 x 9.6 x 5.5 cm (6.06 x 3.78 x 2.17 in.)
Light Source LED
Measurement Technology Absorbance
Plate Type 96-well Plate
Reading Options Endpoint, Kinetic
Shaking None
Software Type PC Software
Voltage 5 V
Weight (English) 1.98 lb.
Weight (Metric) 900 g
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.