Learn More
Vitlab™ Simplex II Bottletop Dispenser
Product Code. 15634854
Change view
Click to view available options
Volume (Metric):
0.2 to 2.0 mL
0.5 to 5.0 mL
1.0 to 10.0 mL
10.0 to 100.0 mL
2.5 to 25 mL
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
15634854 1.0 to 10.0 mL Each
15670915 0.2 to 2.0 mL Each
15624854 0.5 to 5.0 mL Each
15680915 2.5 to 25 mL Each
15600925 10.0 to 100.0 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 15634854 Supplier Vitlab™ Supplier No. 1621505
Only null left
Add to basket
Add to basket
Vitlab™ Simplex II Bottletop Dispenser is designed to be practically universally applicable and can be used with many organic and inorganic solutions.
  • Direct displacement plunger with broad PFA seal prevents crystallisation build-up
  • Settings are quickly and accurately adjusted due to interior scalloped track and practical screw mechanism
  • Screwable ventilation plug for easy connection of a drying tube
  • Screwable discharge valve is equipped with an additional safety bulb and closes the dosing channel, if dispensing tube is not installed, so that no medium can escape
  • Equipped with a positive displacement piston and a fluoroplastic (PFA) sealing lip on the cylinder wall
  • Nominal volume 2 to 10mL includes adapters GL 25, GL 28, GL 32, GL 38, S 40 and telescopic intake tube (length 125 to 240mm)
  • Nominal volume 25 to 100mL includes adapters GL 32, GL 38, S 40 and telescopic intake tube (length 170 to 330mm)
Specifications
Volume (Metric) 1.0 to 10.0 mL