Learn More
Vitlab™ Simplex II Fix Dispenser
Product Code. 15630925
Change view
Click to view available options
Volume (Metric):
1 mL
5 mL
10 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
15630925 10 mL Each
15610925 1 mL Each
15620925 5 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15630925 Supplier Vitlab™ Supplier No. 1622505
In Stock
Add to basket
Add to basket
Vitlab™ Simplex II Fix Dispenser is designed to be practically universally applicable and can be used with many organic and inorganic solutions.
  • Bottle-top dispenser with fixed volume
  • Direct displacement plunger with broad PFA seal prevents crystallisation build-up
  • Screwable ventilation plug for easy connection of a drying tube
  • Screwable discharge valve is equipped with an additional safety bulb and closes the dosing channel, if dispensing tube is not installed, so that no medium can escape
  • Equipped with a positive displacement piston and a fluoroplastic (PFA) sealing lip on the cylinder wall
  • Nominal volume 1 to 10mL includes adapters GL 25, GL 28, GL 32, GL 38, S 40 and telescopic intake tube (length 125 to 240mm)
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 10 mL