missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Simvastatin

Product Code. 30001291
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001291 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30001291 Supplier R&D Systems™ Supplier No. 1965/50
  • ONLINE EXCLUSIVE PRICE
    307.95 €
    275.10 € / 50mg
    Save 32.85 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. HMG-CoA reductase inhibitor

HMG-CoA reductase inhibitor; decreases levels of low density lipoprotein. Has multiple biological effects including bone formation stimulation, inhibition of smooth muscle cell proliferation and migration, induction of ferroptosis, and anticancer and anti-inflammatory activity. Inactive lactone prodrug of simvastatin hydroxy acid, naturally bioactivated in vivo following oral administration.

Deuterated analog also available.

TRUSTED_SUSTAINABILITY

Specifications

CAS 79902-63-9
Quantity 50 mg
Product Line Tocris
Molecular Formula C25H38O5
Formula Weight 418.57
Purity >98%
Molecular Weight (g/mol) 418.574
Chemical Name or Material Simvastatin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.