missing translation for 'onlineSavingsMsg'
Learn More

Simvastatin Hydroxy Acid Sodium Salt, TRC

Product Code. 30003584
Change view
Click to view available options
Quantity:
1 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30003584 25 mg 25mg
30004135 1 mg 1mg
30010240 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30003584 Supplier Toronto Research Chemicals Supplier No. TRCS48505025MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101314-97-0
Molecular Formula C25 H39 O6 . Na
Molecular Weight (g/mol) 458.56
Synonym 1-Naphthaleneheptanoic acid, 8-(2,2-dimethyl-1-oxobutoxy)-1,2,6,7,8,8a-hexahydro-β,δ-dihydroxy-2,6-dimethyl-, sodium salt (1:1), (βR,δR,1S,2S,6R,8S,8aR)-, 1-Naphthaleneheptanoic acid, 8-(2,2-dimethyl-1-oxobutoxy)-1,2,6,7,8,8a-hexahydro-β,δ-dihydroxy-2,6-dimethyl-, monosodium salt, (βR,δR,1S,2S,6R,8S,8aR)- (9CI), 1-Naphthaleneheptanoic acid, 8-(2,2-dimethyl-1-oxobutoxy)-1,2,6,7,8,8a-hexahydro-β,δ-dihydroxy-2,6-dimethyl-, monosodium salt, [1S-[1α(βS*,δS*),2α,6β,8β,8aα]]-, (3R,5R)-7-[(1S,2S,6R,8S,8aR)-8-[(2,2-Dimethylbutanoyl)oxy]-2,6-dimethyl-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoic Acid Sodium Salt, Simvastatin Imp. A (EP) as Sodium Salt, Tenivastatin Sodium Salt, Simvastatin Hydroxy Acid Sodium Salt
IUPAC Name sodium;(3R,5R)-7-[(1S,2S,6R,8S,8aR)-8-(2,2-dimethylbutanoyloxy)-2,6-dimethyl-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoate
SMILES [Na+].CCC(C)(C)C(=O)O[C@H]1C[C@@H](C)C=C2C=C[C@H](C)[C@H](CC[C@@H](O)C[C@@H](O)CC(=O)[O-])[C@@H]12

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C25H40O6.Na/c1-6-25(4,5)24(30)31-21-12-15(2)11-17-8-7-16(3)20(23(17)21)10-9-18(26)13-19(27)14-22(28)29;/h7-8,11,15-16,18-21,23,26-27H,6,9-10,12-14H2,1-5H3,(H,28,29);/q;+1/p-1/t15-,16-,18+,19+,20-,21-,23-;/m0./s1
Formula Weight 458.2644
Percent Purity >95%
Chemical Name or Material (3R,5R)-7-[(1S,2S,6R,8S,8aR)-8-[(2,2-Dimethylbutanoyl)oxy]-2,6-dimethyl-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoic Acid Sodium Salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.