missing translation for 'onlineSavingsMsg'
Learn More

Simvastatin, MedChemExpress

Product Code. 16467336
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16467336 100 mg 100mg
16451266 200 mg 200mg
16433397 50 mg 50mg
16461866 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16467336 Supplier Medchem Express Supplier No. HY17502
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Simvastatin (MK 733) is a competitive inhibitor of HMG-CoA reductase with a Ki of 0.2 nM.

Simvastatin (MK 733) is a competitive inhibitor of HMG-CoA reductase with a Ki of 0.2 nM.

Specifications

Chemical Name or Material Simvastatin
CAS 79902-63-9
Color White
Purity Grade Notes Research
Molecular Formula C25H38O5
Quantity 100 mg
Synonym MK 733
Solubility Information Ethanol : 100 mg/mL (238.91 mM; Need ultrasonic) ∣DMSO : ≥ 50 mg/mL (119.45 mM)
SMILES CCC(C)(C)C(O[C@H]1C[C@@H](C)C=C2C=C[C@H](C)[C@H](CC[C@@H]3C[C@@H](O)CC(O3)=O)[C@@]12[H])=O
Molecular Weight (g/mol) 418.57
Formula Weight 418.57
Percent Purity 99.45%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.