missing translation for 'onlineSavingsMsg'
Learn More

Simvastatin, TRC

Product Code. 30134483
Change view
Click to view available options
Quantity:
25 mg
50 mg
250 mg
Unit Size:
250mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134483 250 mg 250mg
30012366 50 mg 50mg
30004072 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134483 Supplier Toronto Research Chemicals Supplier No. TRCS485000250MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79902-63-9
Molecular Formula C25 H38 O5
Molecular Weight (g/mol) 418.57
Synonym Simvastatin, Butanoic acid, 2,2-dimethyl-, (1S,3R,7S,8S,8aR)-1,2,3,7,8,8a-hexahydro-3,7-dimethyl-8-[2-[(2R,4R)-tetrahydro-4-hydroxy-6-oxo-2H-pyran-2-yl]ethyl]-1-naphthalenyl ester, Butanoic acid, 2,2-dimethyl-, 1,2,3,7,8,8a-hexahydro-3,7-dimethyl-8-[2-(tetrahydro-4-hydroxy-6-oxo-2H-pyran-2-yl)ethyl]-1-naphthalenyl ester, [1S-[1α,3α,7β,8β(2S*,4S*),8aβ]]-, (+)-Simvastatin, Apo-Simva, Apo-Simvastatin, Bestatin 20, Cholestat, Co-Simvastatin, Denan, Egilipid, Eucor, Kolestevan, L 644128-000U, Lipex, Lipinorm, Liponorm, Lipovas, Lodales, Lycostatin, MK 733, Modutrol, Nor-Vastina, Novo-Simvastatin, Pms-simvastatin, Rechol, Simastin 20, Simcovas, Simgal, Simi 20, Simlip, Simlip 20, Simlo 20, Simlup 20, Simovil, Simratio, Simvacard, Simvachol, Simvacor, Simvahexal, Simvas, Simvastatin lactone, Simvasterol, Simvastol, Simvofix, Simvotin, Sinvacor, Sinvascor, Sivastin, Starstat 20, Synvinolin, Valemia, Vasilip, Vastan, Vazilip, Velostatin, Ximve, Zocor, Zocor Forte, Zocord, Zorced, Zosta
IUPAC Name [(1S,3R,7S,8S,8aR)-8-[2-[(2R,4R)-4-hydroxy-6-oxooxan-2-yl]ethyl]-3,7-dimethyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl] 2,2-dimethylbutanoate
SMILES CCC(C)(C)C(=O)O[C@H]1C[C@@H](C)C=C2C=C[C@H](C)[C@H](CC[C@@H]3C[C@@H](O)CC(=O)O3)[C@@H]12

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C25H38O5/c1-6-25(4,5)24(28)30-21-12-15(2)11-17-8-7-16(3)20(23(17)21)10-9-19-13-18(26)14-22(27)29-19/h7-8,11,15-16,18-21,23,26H,6,9-10,12-14H2,1-5H3/t15-,16-,18+,19+,20-,21-,23-/m0/s1
Formula Weight 418.2719
Percent Purity >95%
Chemical Name or Material Simvastatin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.