missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Sinapine thiocyanate

Product Code. 16451506
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mg
Unit Size:
200mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16451506 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16451506 Supplier MedChemExpress Supplier No. HYN0450
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Sinapine thiocyanate is an alkaloid isolated from seeds of the cruciferous species. Sinapine thiocyanate exhibits anti-inflammatory, anti-oxidant, anti-tumor, anti-angiogenic and radio-protective effects. Sinapine thiocyanate is also an acetylcholinesterase (AChE) inhibitor and can be used for the research of Alzheimer’s disease, ataxia, myasthenia gravis, and Parkinson’s disease.

Specifications

Content And Storage 4°C, sealed storage, away from moisture and light∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture and light)
Molecular Weight (g/mol) 368.45 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-programmed cell death
CAS 7431-77-8
Product Type Natural Products
Purity 98.0%
Solubility DMSO : ≥ 125 mg/mL (339.26 mM)
Target P-glycoprotein∣Cholinesterase (ChE)
Quantity 200 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.