missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Single Well High Profile Reagent Reservoirs

Product Code. 11320275 Shop All Axygen Scientific Products
Change view
Click to view available options
Capacity (Metric):
240 mL
282 mL
290 mL
Sterility:
Non-sterile
Sterile
Channels:
8
12
96
384
Unit Size:
Pack of 25
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Sterility Channels Unit Size
11320275 282 mL Sterile 384 Pack of 25
11310275 282 mL Non-sterile 384 Pack of 25
11360275 290 mL Sterile 8 Pack of 25
11300275 290 mL Sterile 12 Pack of 25
11370275 240 mL Non-sterile 96 Pack of 25
11350275 290 mL Non-sterile 8 Pack of 25
11390265 290 mL Non-sterile 12 Pack of 25
11380275 240 mL Sterile 96 Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 11320275 Supplier Axygen™ Supplier No. RESSW384HPSI
  • ONLINE EXCLUSIVE PRICE
    369.00 €
    257.81 € / Pack of 25
    Save 111.19 €
    30% Off
View all promotions
Estimated Shipment: 08-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Manufactured with virgin polypropylene for excellent chemical resistance

Corning offers a complete line of Axygen single-well and multiple-well reagent reservoirs which are available in multiple formats for automated and manual use. All reservoirs are designed to comply with standard ANSI/SLAS footprint dimensions for microplates and are compatible with most automation workstations. Reagent reservoirs are manufactured with polypropylene for excellent chemical resistance.

  • Durable and cost effective
  • Various formats for multiple applications
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ANSI/SLAS, RNase/DNase-free, non-pyrogenic
Color Clear
Capacity (Metric) 282 mL
Material Polypropylene
Sterility Sterile
Type Reagent Reservoir
Packaging Individually Wrapped; 25/Cs.
Channels 384
Disposable Single-use
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.