missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Single Well Low Profile Reagent Reservoirs

Product Code. 11370265 Shop All Axygen Scientific Products
Change view
Click to view available options
Capacity (Metric):
86 mL
90 mL
92 mL
Sterility:
Non-sterile
Sterile
Channels:
1
96
384
Packaging:
25/Cs.
Individually Wrapped; 25/Cs.
Unit Size:
Pack of 25
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Sterility Channels Packaging Unit Size
11370265 90 mL Non-sterile 1 25/Cs. Pack of 25
11301974 86 mL Non-sterile 96 25/Cs. Pack of 25
11330275 92 mL Non-sterile 384 25/Cs. Pack of 25
11340275 92 mL Sterile 384 Individually Wrapped; 25/Cs. Pack of 25
11380265 90 mL Sterile 1 Individually Wrapped; 25/Cs. Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11370265 Supplier Axygen™ Supplier No. RESSW1LP
  • ONLINE EXCLUSIVE PRICE
    364.00 €
    254.51 € / Pack of 25
    Save 109.49 €
    30% Off
View all promotions
Estimated Shipment: 09-04-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Manufactured with virgin polypropylene for excellent chemical resistance

Corning offers a complete line of Axygen single-well and multiple-well reagent reservoirs which are available in multiple formats for automated and manual use. All reservoirs are designed to comply with standard ANSI/SLAS footprint dimensions for microplates and are compatible with most automation workstations. Reagent reservoirs are manufactured with polypropylene for excellent chemical resistance.

  • Durable and cost effective
  • Various formats for multiple applications
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ANSI/SLAS, RNase/DNase-free, non-pyrogenic
Color Clear
Capacity (Metric) 90 mL
Material Polypropylene
Sterility Non-sterile
Type Reagent Reservoir
No. per Case 25/Cs.
No. per Pack 25/Pk.
Packaging 25/Cs.
Channels 1
Disposable Single-use
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.