missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Single Well Reagent Reservoir with 96-Well Bottom Troughs, High Profile, Sterile

Product Code. 30306807 Shop All Axygen Scientific Products
Change view
Click to view available options
Quantity:
25/Cs.
Unit Size:
Pack of 25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30306807 25/Cs. Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30306807 Supplier Axygen™ Supplier No. RESSW96HPSIM
  • ONLINE EXCLUSIVE PRICE
    407.00 €
    358.57 € / Pack of 25
    Save 48.43 €
    11% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

96-well reservoir designed to allow collection pools at the bottom surface to help minimize dead volume.

  • The Axygen brand offers a complete line of single well and multiple well reagent reservoirs which are available in numerous formats for automated and manual applications
  • All reservoirs are designed to comply with ANSI/SLAS 1-2004: Microplates - Footprint Dimensions and are compatible with most automation systems
  • Manufactured with virgin polypropylene for excellent chemical resistance
  • Diamond Shaped Well Series: 96 or 384 well collection pools at the bottom surface help minimize dead volume, RNase-/DNase-free, Nonpyrogenic, Sterile
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Capacity (Metric) 240 mL
Material Polypropylene
Sterility Sterile
Type Reagent Reservoir
For Use With (Application) Hold and Distribute Liquids
Quantity 25/Cs.
Autoclavable Not Autoclavable
Channels 1
Disposable Single-use
Bottom Shape Flat
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.