missing translation for 'onlineSavingsMsg'
Learn More

Berthold Technologies Sirius 2 injection Sample Holder/Reflector For Microfuge Vials

Product Code. 31777424 Shop All Berthold Technologies Products
Change view
Click to view available options
For Use With (Application):
Luminescence Assays
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) Unit Size
31777424 Luminescence Assays Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31777424 Supplier Berthold Technologies Supplier No. 84720
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Optional sample holder for measuring and injecting in microcentrifuge tubes.

  • Standard sample holder supports luminescence tubes (12 mm diameter, 55 mm or 75 mm length)
  • Not compatible with microcentrifuge tubes
  • Optional sample holder enables measurement and injection in microcentrifuge tubes

Specifications

Type Adapter
Color Black
For Use With (Application) Luminescence Assays
For Use With (Equipment) Sirius 2 LB 9526 Tube Luminometer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.