missing translation for 'onlineSavingsMsg'
Learn More

Berthold Technologies Sirius 2 LB 9526 Tube Luminometer

Product Code. 31777398 Shop All Berthold Technologies Products
Change view
Click to view available options
Includes:
1 Injector
2 Injector
Without Injector
Connection Type:
PC Operation
Touchscreen Operation
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Connection Type Unit Size
31777398 1 Injector Touchscreen Operation Each
31777324 Without Injector Touchscreen Operation Each
31777397 2 Injector Touchscreen Operation Each
31777310 Without Injector PC Operation Each
31777298 1 Injector PC Operation Each
31777348 2 Injector PC Operation Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 31777398 Supplier Berthold Technologies Supplier No. 8401802
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Compact, sensitive tube luminometer for flash assays with upgrade options and stand-alone operation.

High-performance luminescence detection

  • Detects less than 0.5 zmol of firefly luciferase (1 amol ATP/tube)
  • Light collection maximized through close proximity of the sample to the detector and optimized light reflectors surrounding the sample vial
  • The detector operated in photon counting mode, ensuring very low background signal, excellent signal-to-noise ratio, and the highest linearity

Outstanding flexibility

  • Sample chamber designed for performance and flexibility - pick the sample format that fits your application best
  • Optional sample holders enable accurate injection into microcentrifuge tubes and standard luminescence tubes
  • Compatible with standard 12 mm luminescence tubes (55 and 75 mm) and microcentrifuge tubes (1.5 and 2 mL)

Intuitive touchscreen operation

  • Touchscreen-driven software operated by touch gestures
  • Easy-to-use software with advanced features such as handling of replicates, dual assay calculations, and display of graphics
  • Data can be downloaded onto a USB stick, printed, or exported for further calculation in Excel or other programs

Flexible injector solutions

  • Available with up to two syringe-type automatic reagent injectors
  • Injection volume range of 20 to 500 μL
  • Ready-to-go sample holders for precise injection for 1.5 mL and 2 mL microfuge tubes, as well as 55 mm and 75 mm, 12 mm diameter tubes
  • Includes: Luminometer, Power Brick, User Manuals, Standard Accessories

Specifications

Type Luminometer
Display Type 7 in. Capacitive Touch, Gesture Control, 480 x 800 Pixels
Connection Type Touchscreen Operation
Detector Low-noise Photomultiplier Tube in Single Photon Counting Mode
Detection Method Luminescence
Detection Limits <1 amol ATP
Volume (Metric) Dispense 20 to 500 μL
Color Light Gray
For Use With (Application) Luminescence Assays
Voltage 24 V
Frequency 50/60 Hz
For Use With (Equipment) Tubes
Height (Metric) 241 mm
Width (Metric) 349 mm
Depth (Metric) 150 mm
Weight (Metric) 7.5 kg
Includes 1 Injector
Storage Requirements 0°C to 40°C
Certifications/Compliance CE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.