missing translation for 'onlineSavingsMsg'
Learn More

Berthold Technologies Sirius 2 Sample Holder/Reflector For Microfuge Vials

Product Code. 31777406 Shop All Berthold Technologies Products
Change view
Click to view available options
For Use With (Application):
Luminescence Applications
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) Unit Size
31777406 Luminescence Applications Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31777406 Supplier Berthold Technologies Supplier No. 84722
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Optional sample holder for measuring in microcentrifuge tubes.

  • The standard sample holder supports luminescence tubes (12 mm diameter, 55 mm or 75 mm length)
  • Not compatible with microcentrifuge tubes
  • The optional sample holder enables measurement only (no injection) in microcentrifuge tubes
  • Injection in microcentrifuge tubes requires sample holder 84720

Specifications

Type Adapter
Color Black
For Use With (Application) Luminescence Applications
For Use With (Equipment) Sirius 2 LB 9526 Tube Luminometer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.