missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SirReal 2

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
SirReal 2 is a selective SIRT2 inhibitor (IC50 = 140 nM). Has minimal effect on SIRT1 and SIRT3-6. Increases α-tubulin acetylation levels in HeLa cells.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | SirReal 2 |
| Purity | >98% |
| Molecular Weight (g/mol) | 420.55 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction