missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Sirtinol

Product Code. 17120282
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17120282 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17120282 Supplier STEMCELL Technologies Supplier No. 73824
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Epigenetic modifier; Inhibitor of Sir2p, SIRT1, and SIRT2

  • Sirtinol is a cell-permeable inhibitor of sirtuin NAD+-dependent histone deacetylases (HDACs), including yeast Sir2p (IC50 = 68 μM; Grozinger et al.), human SIRT1 (IC50 = 131 μM; Schuetz et al.), and SIRT2 (IC50 = 38 μM; Grozinger et al.) It does not inhibit human HDAC1 activity (Grozinger et al.) Sirtinol also activates the auxin signaling pathway in Arabidopsis (Dai et al.)
  • Suppresses inflammatory signaling in human dermal microvascular endothelial cells (Orecchia et al.)
  • Enhances apoptosis and cell death in leukemic cells, but not in healthy leukocytes and hematopoietic progenitors (Cea et al.)
  • Induces senescence-like growth arrest in human cancer cell lines MCF-7 and H1299 (Ota et al.; Wang et al.)

Specifications

CAS 410536-97-9
Synonym Sir Two Inhibitor Naphthol
Quantity 10 mg
Molecular Formula C26H22N2O2
Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.