missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies SIS3

Product Code. 17130302
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
500 μg
Unit Size:
1mg
500µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17130302 1 mg 1mg
17120302 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17130302 Supplier STEMCELL Technologies Supplier No. 73964
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Activin/NODAL/TGF-β pathway inhibitor; Inhibits SMAD3

  • SIS3 (specific inhibitor of SMAD3) is a cell-permeable selective inhibitor of transforming growth factor β1 (TGF-β1)-dependent SMAD3 phosphorylation (IC50 = 3 μM) and SMAD3-mediated cellular signaling (Jinnin et al.) SIS3 has no effect on SMAD2, p38 mitogen-activated protein kinase (MAPK), extracellular signal-regulated kinase (ERK), or phosphoinositide 3-kinases
  • Facilitates the production of anterior ectoderm from mouse induced pluripotent stem cells (Oshima et al.)
  • Inhibits myofibroblast differentiation of normal human dermal fibroblasts (Jinnin et al.)

Specifications

CAS 1009104-85-1
Synonym Smad3 inhibitor
Quantity 1 mg
Molecular Formula C28H27N3O3
Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.