missing translation for 'onlineSavingsMsg'
Learn More

Sisomicin Sulfate Salt, TRC

Product Code. 30134122
Change view
Click to view available options
Quantity:
100 mg
500 mg
5 g
Unit Size:
100mg
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134122 500 mg 500mg
30136679 5 g 5g
17939849 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134122 Supplier Toronto Research Chemicals Supplier No. TRCS488505500MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53179-09-2
Molecular Formula 2 C19 H37 N5 O7 . 5 H2 O4 S
Molecular Weight (g/mol) 1385.45
Synonym D-Streptamine, O-3-deoxy-4-C-methyl-3-(methylamino)-β-L-arabinopyranosyl-(1→6)-O-[2,6-diamino-2,3,4,6-tetradeoxy-α-D-glycero-hex-4-enopyranosyl-(1→4)]-2-deoxy-, sulfate (2:5), D-Streptamine, O-3-deoxy-4-C-methyl-3-(methylamino)-β-L-arabinopyranosyl-(1→6)-O-[2,6-diamino-2,3,4,6-tetradeoxy-α-D-glycero-hex-4-enopyranosyl-(1→4)]-2-deoxy-, sulfate (2:5) (salt) (9CI), Antibiotic 66-40 sulfate, Baymicin, Extramycin, Mensiso, Sisobiotic, Sisolline, Sisomicin sulfate, Sisomin, Sisomycin sulfate, Sissomicin sulfate
IUPAC Name (2R,3R,4R,5R)-2-[(1S,2S,3R,4S,6R)-4,6-diamino-3-[[(2S,3R)-3-amino-6-(aminomethyl)-3,4-dihydro-2H-pyran-2-yl]oxy]-2-hydroxycyclohexyl]oxy-5-methyl-4-(methylamino)oxane-3,5-diol;sulfuric acid
SMILES CN[C@@H]1[C@@H](O)[C@@H](O[C@H]2[C@H](N)C[C@H](N)[C@@H](O[C@H]3OC(=CC[C@H]3N)CN)[C@@H]2O)OC[C@]1(C)O.CN[C@@H]4[C@@H](O)[C@@H](O[C@H]5[C@H](N)C[C@H](N)[C@@H](O[C@H]6OC(=CC[C@H]6N)CN)[C@@H]5O)OC[C@]4(C)O.OS(=O)(=O)O.OS(=O)(=O)O.OS(=O)(=O)O.OS(=O)(=O)O.OS(=O)(=O)O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/2C19H37N5O7.5H2O4S/c2*1-19(27)7-28-18(13(26)16(19)24-2)31-15-11(23)5-10(22)14(12(15)25)30-17-9(21)4-3-8(6-20)29-17;5*1-5(2,3)4/h2*3,9-18,24-27H,4-7,20-23H2,1-2H3;5*(H2,1,2,3,4)/t2*9-,10+,11-,12+,13-,14-,15+,16-,17-,18-,19+;;;;;/m11...../s1
Formula Weight 1384.3755
Percent Purity >95%
Chemical Name or Material Sisomicin Sulfate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.