missing translation for 'onlineSavingsMsg'
Learn More

Phytosterols, Practical Grade, MP Biomedicals™

Product Code. 11406272 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11406272 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11406272 Supplier MP Biomedicals Supplier No. 0210288690
 Restricted Item
Estimated Shipment: 08-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 83-46-5
Molecular Formula C29H50O
Molecular Weight (g/mol) 414.718
InChI Key KZJWDPNRJALLNS-VJSFXXLFSA-N
Synonym 22,23-dihydrostigmasterol, α-Dihydrofucosterol, 24β-Ethylcholesterol, 5-Stigmasten-3β-ol, 22,23-dihydrostigmasterol, azuprostat, triastonal, cinchol, rhamnol, harzol
PubChem CID 222284
ChEBI CHEBI:27693
IUPAC Name (3S,8S,9S,10R,13R,14S,17R)-17-[(2R,5R)-5-ethyl-6-methylheptan-2-yl]-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
SMILES CCC(CCC(C)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)O)C)C)C(C)C

Specifications

Color White
Quantity 500 g
Formula Weight 414.7
Grade Practical
Physical Form Powder
Chemical Name or Material Phytosterols
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.