missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SK 575-NEG
GREENER_CHOICE

Product Code. 18767636
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18767636 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18767636 Supplier Tocris Bioscience™ Supplier No. 7588/5
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Negative control for SK 575 (Cat. No. 7583)

SK 575-NEG is the negative control for SK 575 (Cat. No. 7583).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 12-(4-(2-Fluoro-5-((4-oxo-3,4-dihydrophthalazin-1-yl)methyl)benzoyl)piperazin-1-yl)-N-(2-((2-(1-methyl-2,6-dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)amino)ethyl)-12-oxododecanamide
CAS 2523017-04-9
Quantity 5 mg
Target Active Degrader Negative Controls
Molecular Formula C48H55FN8O8
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.