missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SK 575
GREENER_CHOICE

Product Code. 18788436
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18788436 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18788436 Supplier Tocris Bioscience™ Supplier No. 7583/5
Stock Up and Save This Summer
Use promo code 27950
View all promotions
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Potent PARP1 Degrader (PROTAC™)

SK 575 is a potent PARP1 Degrader (PROTAC®; DC50 ≤ 6.72 nM, >99% PARP1 degradation at 10 nM). It comprises the PARP1/2 inhibitor Olaparib (Cat. No. 7579), joined by a linker to cereblon/cullin 4A ligand Thalidomide (Cat. No. 0652). SK 575 inhibits the growth of various cancer cell lines bearing BRCA1/2 mutations with IC50 values in the nanomolar range. It effectively reduces PARP1 protein levels in mouse SW620 tumor xenograft models. SK 575 dose-dependently potentiates the antitumor activity of Temozolomide (Cat. No. 2706) and Cisplatin (Cat. No. 2251) in vivo.

PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(2-((2-(2,6-Dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)amino)ethyl)-12-(4-(2-fluoro-5-((4-oxo-3,4-dihydrophthalazin-1-yl)methyl)benzoyl)piperazin-1-yl)-12-oxododecanamide
CAS 2523016-96-6
Quantity 5 mg
Target PARP
Molecular Formula C47H53FN8O8
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.