missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ SKF 81297 hydrobromide

Product Code. 18065032
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18065032 50 mg 50mg
18024162 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18065032 Supplier R&D Systems™ Supplier No. 1447/50
  • ONLINE EXCLUSIVE PRICE
    1210.00 €
    1142.40 € / 50mg
    Save 67.60 €
    5% Off
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. D1 agonist

Dopamine D1-like receptor agonist. Centrally active following systemic administration in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67287-39-2
Molecular Formula C16H17BrClNO2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 370.67
Purity >98%
Molecular Weight (g/mol) 370.671
Chemical Name or Material SKF 81297 hydrobromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.