missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ SKF 96365 hydrochloride

Product Code. 18085252
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18085252 10 mg 10mg
18045652 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18085252 Supplier R&D Systems™ Supplier No. 1147/10
  • ONLINE EXCLUSIVE PRICE
    307.95 €
    276.15 € / 10mg
    Save 31.80 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. TRPC channel blocker; also inhibits store-operated Ca2+ entry

Store-operated Ca2+ entry (SOCE) inhibitor that inhibits STIM1. Also blocks TRPC channels, voltage-gated Ca2+ channels and potassium channels.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130495-35-1
Molecular Formula C22H27ClN2O3

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 402.92
Purity >99%
Molecular Weight (g/mol) 402.919
Chemical Name or Material SKF 96365 hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.