missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals SKF-96365 hydrochloride, 99+%

Product Code. 15483619
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15483619 10 mg 10mg
15493619 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15483619 Supplier Thermo Scientific Chemicals Supplier No. J60937.MA
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This item is not returnable. View return policy

Inhibits receptor-mediated calcium influx

Inhibits receptor-mediated calcium influx

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130495-35-1
Molecular Formula C22H26N2O3·HCl, C22H27ClN2O3
Synonym 1-[2-(4-Methoxyphenyl)-2-[3-(4-methoxyphenyl)propoxy]ethyl]imidazole

Specifications

Quantity 10 mg
Description White
For Use With (Application) Inhibits receptor-mediated calcium influx
Formula Weight 402.92g/mol
Solubility Soluble to 100mM in water
Product Type Signal Transduction Reagent
Form Powder
InChI Key FWLPKVQUECFKSW-UHFFFAOYNA-N
SMILES [H+].[Cl-].COC1=CC=C(CCCOC(CN2C=CN=C2)C2=CC=C(OC)C=C2)C=C1
IUPAC Name hydrogen 1-[2-(4-methoxyphenyl)-2-[3-(4-methoxyphenyl)propoxy]ethyl]-1H-imidazole chloride
Molecular Weight (g/mol) 402.92
Assay ≥99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.