Learn More
Thermo Scientific™ Skylite Refilling Reference Electrolyte for Eutech™ EC620130 pH electrode
Description
The measure of a liquid's acidity and alkalinity, pH is one of the most common parameter measured in a wide variety of industries ranging from water and wastewater treatment, chemical production, agriculture research and production, environmental monitoring, chemical and life sciences research, biochemical and pharmaceutical research, electronics production to food processing and other industrial applications.
Eutech™ offers a wide range of pH buffers, calibration, and reference solutions for use with pH meters and electrodes.
Features:
- Buffers and calibration solutions packed in 60 mL durable plastic bottles – economical and convenient for laboratory use.
Specifications
Specifications
| Capacity (Metric) | 60 mL |
| Color | Light Blue |
| Description | Refilling reference electrolyte for pH electrode |
| For Use With (Equipment) | EC620130 pH Electrode |
| Type | Reference Solution |
| Melting Point | ∼100°C |
| Odor | Odorless |
| pH Range | 7 |
| Solubility | Soluble |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.