missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SL 0101-1
GREENER_CHOICE

Product Code. 18093792
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18093792 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18093792 Supplier Tocris Bioscience™ Supplier No. 2250/1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective ribosomal S6 kinase (RSK) inhibitor

Selective inhibitor of ribosomal S6 kinase (RSK) (IC50 = 89 nM for RSK2). Does not inhibit upstream kinases such as MEK, Raf and PKC. Inhibits the growth of MCF-7 human breast cancer cells with no effect on the normal breast cell line.
TRUSTED_SUSTAINABILITY

Specifications

CAS 77307-50-7
Quantity 1 mg
Molecular Formula C25H24O12
Formula Weight 516.46
Purity >98%
Molecular Weight (g/mol) 516.455
Chemical Name or Material SL 0101-1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.