missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC25A15 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
196.00 € - 468.00 €
Specifications
| Antigen | SLC25A15 |
|---|---|
| Dilution | Western Blot 1:200-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
SLC25A15 Polyclonal antibody specifically detects SLC25A15 in Human samples. It is validated for Western BlotSpecifications
| SLC25A15 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| D13S327, mitochondrial ornithine transporter 1, ORC1, ornithine transporter 1, ORNT1HHH, solute carrier family 25 (mitochondrial carrier; ornithine transporter) member15, Solute carrier family 25 member 15 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-301 of human SLC25A15 (NP_055067.1). MKSNPAIQAAIDLTAGAAGGTACVLTGQPFDTMKVKMQTFPDLYRGLTDCCLKTYSQVGFRGFYKGTSPALIANIAENSVLFMCYGFCQQVVRKVAGLDKQAKLSDLQNAAAGSFASAFAALVLCPTELVKCRLQTMYEMETSGKIAKSQNTVWSVIKSILRKDGPLGFYHGLSSTLLREVPGYFFFFGGYELSRSFFASGRSKDELGPVPLMLSGGVGGICLWLAVYPVDCIKSRIQVLSMSGKQAGFIRTFINVVKNEGITALYSGLKPTMIRAFPANGALFLAYEYSRKLMMNQLEAY | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:200-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 10166 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title