missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sampling Systems™ Sleeve Sample Chambers
Quickly attached to the same handle to allow different sample volumes to be taken. Sample Systems™ Sleeve Sample Chambers are ideal for sampling large volumes at great depths.
Brand: Sampling Systems™ 1035T-100
This item is not returnable.
View return policy
Description
- Made from high quality 316L stainless steel
- Bright finish
- Single Point
- Easy to use
Specifications
Sample Chamber | |
Stainless Steel (316L) | |
100 mL | |
Powder Sampling |