Learn More
ibidi μ-Slide 1 Well Untreated
Product Code. 31941050
Encompass_Preferred
Change view
Click to view available options
No. per Case:
120/Cs.
15/Cs.
Unit Size:
Pack of 120
Pack of 15
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. per Case Unit Size
31941050 120/Cs. Pack of 120
30130591 15/Cs. Pack of 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 31941050 Supplier ibidi Supplier No. 82101120
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
A μ-Slide for cell culture and high-end microscopy.
  • No. 1.5 polymer coverslip
  • Hydrophobic
  • Sterilized
  • Individually packed
Specifications
Type Chamber Slide
Surface Treatment Untreated
Material Polymer
For Use With (Application) Cell Culture, High-end Microscopy
No. per Case 120/Cs.
No. per Pack 8/Tray, 15 Trays
Sterility Sterile