Learn More
Ibidi μ-Slide 18 Well - Flat ibiTreat
Click to view available options
:
μ-Slide
Description
- A chambered coverslip with 18 wells for use in matrix tests and quick immunofluorescence stainings
- bottom material: #1.5 Polymer Coverslip
- tissue culture treated
- sterilized
Specifications
Specifications
| Type | Chambered Coverglass |
| Sterility | Sterile |